Assay Details
| Gene Symbol: NF-YA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336524.1 | NP_001323828.1 | 3 - 4 | 802 | 65 | |
| NM_001336525.1 | NP_001323827.1 | 1 - 2 | 339 | 65 | ||
| NM_179904.5 | NP_850235.1 | 4 - 5 | 417 | 65 | ||
| GenBank mRNA | AY072414.1 | - | 3 - 4 | 278 | 65 | |
| AY114711.1 | - | 3 - 4 | 225 | 65 | ||
More Information
Set Membership: |
|

NF-YA4
Genomic Map