Assay Details
Gene Symbol: TRFL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036420.2 | NP_001031497.1 | 1 - 2 | 899 | 130 | |
NM_001336621.1 | NP_001324498.1 | 2 - 3 | 197 | 130 | ||
NM_179944.3 | NP_850275.1 | 2 - 3 | 275 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
