Assay Details
Gene Symbol: TRFL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036420.2 | NP_001031497.1 | 3 - 4 | 1140 | 70 | |
NM_001336621.1 | NP_001324498.1 | 4 - 5 | 438 | 70 | ||
NM_179944.3 | NP_850275.1 | 4 - 5 | 516 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
