Assay Details
Gene Symbol: TRFL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036420.2 | NP_001031497.1 | 5 - 6 | 1511 | 87 | |
NM_001336621.1 | NP_001324498.1 | 6 - 7 | 809 | 87 | ||
NM_179944.3 | NP_850275.1 | 6 - 7 | 887 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
