Assay Details
Gene Symbol: LCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036429.2 | NP_001031506.1 | 2 - 3 | 785 | 85 | |
NM_001336677.1 | NP_001324059.1 | 1 - 2 | 940 | 85 | ||
NM_179956.2 | NP_850287.1 | 2 - 3 | 691 | 85 | ||
NM_179957.3 | NP_850288.2 | 1 - 2 | 898 | 85 | ||
GenBank mRNA | AK316808.1 | - | 2 - 3 | 682 | 85 | |
AY081345.1 | - | 2 - 3 | 681 | 85 | ||
BT002153.1 | - | 1 - 2 | 576 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
