Assay Details
| Gene Symbol: FAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129384.3 | NP_565886.1 | 17 - 18 | 2530 | 85 | |
| NM_179963.3 | NP_850294.1 | 17 - 18 | 2530 | 85 | ||
| GenBank mRNA | AY056301.1 | - | 17 - 18 | 2434 | 85 | |
| AY133852.1 | - | 17 - 18 | 2351 | 85 | ||
More Information
Set Membership: |
|

FAC1
Genomic Map