Assay Details
| Gene Symbol: EMB2765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179966.2 | NP_850297.1 | 7 - 8 | 1355 | 69 | |
| GenBank mRNA | AY099864.1 | - | 7 - 8 | 1265 | 69 | |
More Information
Set Membership: |
|

EMB2765
Genomic Map