Assay Details
| Gene Symbol: NF-YB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036433.2 | NP_001031510.1 | 2 - 3 | 297 | 100 | |
| NM_001036434.2 | NP_001031511.1 | 2 - 3 | 388 | 100 | ||
| NM_001202775.1 | NP_001189704.1 | 2 - 3 | 300 | 100 | ||
| NM_001202776.2 | NP_001189705.1 | 2 - 3 | 297 | 100 | ||
| NM_001336741.1 | NP_001323549.1 | 2 - 3 | 342 | 100 | ||
| NM_001336742.1 | NP_001323550.1 | 2 - 3 | 297 | 100 | ||
| NM_001336743.1 | NP_001318379.1 | 2 - 3 | 388 | 100 | ||
| NM_001336744.1 | NP_001323548.1 | 2 - 3 | 388 | 100 | ||
| NM_129445.3 | NP_030436.1 | 2 - 3 | 388 | 100 | ||
| NM_179973.4 | NP_850304.2 | 2 - 3 | 388 | 100 | ||
| NM_179974.4 | NP_850305.2 | 2 - 3 | 388 | 100 | ||
| GenBank mRNA | AK318866.1 | - | 2 - 3 | 277 | 100 | |
| AY088554.1 | - | 2 - 3 | 314 | 100 | ||
| BT004266.1 | - | 2 - 3 | 277 | 100 | ||
| BT005536.1 | - | 1 - 2 | 196 | 100 | ||
| DQ333305.1 | - | 2 - 3 | 297 | 100 | ||
| Y13723.1 | - | 298 | 100 | |||
More Information
Set Membership: |
|

NF-YB1
Genomic Map