Assay Details
| Gene Symbol: NF-YB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036433.2 | NP_001031510.1 | 4 - 5 | 517 | 107 | |
| NM_001036434.2 | NP_001031511.1 | 5 - 6 | 610 | 107 | ||
| NM_001202775.1 | NP_001189704.1 | 4 - 5 | 516 | 107 | ||
| NM_001202776.2 | NP_001189705.1 | 5 - 6 | 588 | 107 | ||
| NM_001336743.1 | NP_001318379.1 | 4 - 5 | 944 | 107 | ||
| NM_179973.4 | NP_850304.2 | 5 - 6 | 610 | 107 | ||
| GenBank mRNA | BT004266.1 | - | 5 - 6 | 499 | 107 | |
More Information
Set Membership: |
|

NF-YB1
Genomic Map