Assay Details
| Gene Symbol: NF-YB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036434.2 | NP_001031511.1 | 6 - 7 | 725 | 99 | |
| NM_001202776.2 | NP_001189705.1 | 6 - 7 | 703 | 99 | ||
| NM_179974.4 | NP_850305.2 | 5 - 6 | 813 | 99 | ||
| GenBank mRNA | BT004266.1 | - | 6 - 7 | 614 | 99 | |
More Information
Set Membership: |
|

NF-YB1
Genomic Map