Assay Details
Gene Symbol: APC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036436.1 | NP_001031513.1 | 11 - 12 | 876 | 109 | |
NM_179978.3 | NP_850309.1 | 11 - 12 | 966 | 109 | ||
GenBank mRNA | AY072346.1 | - | 11 - 12 | 876 | 109 | |
BT002547.1 | - | 11 - 12 | 809 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
