Assay Details
Gene Symbol: PIP1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083972.1 | NP_001077441.1 | 2 - 3 | 427 | 64 | |
NM_100044.5 | NP_171668.1 | 1 - 2 | 697 | 64 | ||
GenBank mRNA | AF348574.1 | - | 1 - 2 | 330 | 64 | |
X69294.1 | - | 343 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
