Assay Details
| Gene Symbol: SPPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035853.1 | NP_001030930.1 | 12 - 13 | 1756 | 64 | |
| NM_001331286.1 | NP_001318903.1 | 12 - 13 | 2036 | 64 | ||
| NM_001331287.1 | NP_001323100.1 | 12 - 13 | 2040 | 64 | ||
| NM_100047.4 | NP_171671.2 | 13 - 14 | 1764 | 64 | ||
| GenBank mRNA | AK229822.1 | - | 13 - 14 | 1613 | 64 | |
More Information
Set Membership: |
|

SPPL4
Genomic Map