Assay Details
| Gene Symbol: ROPGEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331294.1 | NP_001321886.1 | 2 - 3 | 653 | 92 | |
| NM_001331295.1 | NP_001321887.1 | 2 - 3 | 460 | 92 | ||
| NM_001331296.1 | NP_001321884.1 | 2 - 3 | 601 | 92 | ||
| NM_001331297.1 | NP_001321885.1 | 2 - 3 | 653 | 92 | ||
| NM_100052.2 | NP_171676.1 | 2 - 3 | 653 | 92 | ||
More Information
Set Membership: |
|

ROPGEF2
Genomic Map