Assay Details
| Gene Symbol: ROPGEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331295.1 | NP_001321887.1 | 5 - 6 | 1019 | 102 | |
| NM_001331296.1 | NP_001321884.1 | 4 - 5 | 1229 | 102 | ||
| NM_100052.2 | NP_171676.1 | 5 - 6 | 1191 | 102 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Detect Genomic DNA  Arabidopsis thaliana  Probe spans exons | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 ROPGEF2
ROPGEF2 Genomic Map
Genomic Map Back To Top
 Back To Top