Assay Details
Gene Symbol: SEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197959.1 | NP_001184888.1 | 12 - 13 | 1105 | 111 | |
NM_001331318.1 | NP_001320381.1 | 12 - 13 | 1274 | 111 | ||
NM_001331319.1 | NP_001320383.1 | 11 - 12 | 1345 | 111 | ||
NM_001331320.1 | NP_001320382.1 | 11 - 12 | 1339 | 111 | ||
NM_001331321.1 | NP_001320384.1 | 6 - 7 | 633 | 111 | ||
NM_100081.5 | NP_563643.4 | 12 - 13 | 1466 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
