Assay Details
Gene Symbol: SEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197959.1 | NP_001184888.1 | 16 - 17 | 1548 | 74 | |
NM_001331318.1 | NP_001320381.1 | 16 - 17 | 1717 | 74 | ||
NM_001331319.1 | NP_001320383.1 | 15 - 16 | 1788 | 74 | ||
NM_001331320.1 | NP_001320382.1 | 15 - 16 | 1782 | 74 | ||
NM_001331321.1 | NP_001320384.1 | 10 - 11 | 1076 | 74 | ||
NM_100081.5 | NP_563643.4 | 16 - 17 | 1909 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
