Assay Details
Gene Symbol: SEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197959.1 | NP_001184888.1 | 2 - 3 | 211 | 112 | |
NM_001331318.1 | NP_001320381.1 | 2 - 3 | 380 | 112 | ||
NM_001331319.1 | NP_001320383.1 | 2 - 3 | 361 | 112 | ||
NM_100081.5 | NP_563643.4 | 2 - 3 | 572 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
