Assay Details
Gene Symbol: SEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197959.1 | NP_001184888.1 | 10 - 11 | 936 | 77 | |
NM_001331318.1 | NP_001320381.1 | 10 - 11 | 1105 | 77 | ||
NM_001331319.1 | NP_001320383.1 | 9 - 10 | 1176 | 77 | ||
NM_001331320.1 | NP_001320382.1 | 9 - 10 | 1170 | 77 | ||
NM_001331321.1 | NP_001320384.1 | 4 - 5 | 464 | 77 | ||
NM_100081.5 | NP_563643.4 | 10 - 11 | 1297 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
