Assay Details
| Gene Symbol: MAGO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100094.3 | NP_171716.1 | 2 - 3 | 447 | 80 | |
| GenBank mRNA | AY065157.1 | - | 2 - 3 | 430 | 80 | |
| AY087464.1 | - | 2 - 3 | 430 | 80 | ||
| AY114602.1 | - | 2 - 3 | 392 | 80 | ||
| KF051000.1 | - | 2 - 3 | 392 | 80 | ||
More Information
Set Membership: |
|

MAGO
Genomic Map