Assay Details
| Gene Symbol: HFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331332.1 | NP_001323315.1 | 1 - 2 | 689 | 127 | |
| NM_100115.3 | NP_563650.1 | 2 - 3 | 565 | 127 | ||
| GenBank mRNA | AF288287.1 | - | 1 - 2 | 370 | 127 | |
| AF323182.1 | - | 2 - 3 | 529 | 127 | ||
| AF324245.1 | - | 2 - 3 | 444 | 127 | ||
More Information
Set Membership: |
|

HFR1
Genomic Map