Assay Details
| Gene Symbol: AT1G02460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100127.3 | NP_563654.1 | 5 - 6 | 1134 | 99 | |
| GenBank mRNA | BT023462.1 | - | 5 - 6 | 1015 | 99 | |
More Information
Set Membership: |
|

AT1G02460
Genomic Map