Assay Details
Gene Symbol: WEE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100178.3 | NP_171796.1 | 4 - 5 | 793 | 66 | |
GenBank mRNA | AB077385.1 | - | 4 - 5 | 715 | 66 | |
AJ439593.1 | - | 4 - 5 | 793 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
