Assay Details
Gene Symbol: BSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331412.1 | NP_001321215.1 | 14 - 15 | 1474 | 77 | |
NM_100227.6 | NP_171844.6 | 14 - 15 | 1474 | 77 | ||
GenBank mRNA | AY372269.1 | - | 14 - 15 | 1474 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
