Assay Details
Gene Symbol: BSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100227.6 | NP_171844.6 | 21 - 22 | 2429 | 79 | |
GenBank mRNA | AY372269.1 | - | 21 - 22 | 2429 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
