Assay Details
Gene Symbol: BSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331412.1 | NP_001321215.1 | 5 - 6 | 610 | 70 | |
NM_100227.6 | NP_171844.6 | 5 - 6 | 610 | 70 | ||
GenBank mRNA | AY372269.1 | - | 5 - 6 | 610 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
