Assay Details
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035874.1 | NP_001030951.1 | 2 - 3 | 545 | 110 | |
NM_100263.4 | NP_171880.1 | 2 - 3 | 827 | 110 | ||
GenBank mRNA | AB493428.1 | - | 2 - 3 | 551 | 110 | |
AJ630476.1 | - | 2 - 3 | 545 | 110 | ||
AK227837.1 | - | 2 - 3 | 624 | 110 | ||
AY568648.1 | - | 2 - 3 | 545 | 110 | ||
BT006209.1 | - | 2 - 3 | 551 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
