Assay Details
Gene Symbol: VPS2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100276.4 | NP_563696.1 | 10 - 11 | 739 | 65 | |
GenBank mRNA | AY052238.1 | - | 10 - 11 | 734 | 65 | |
AY060501.1 | - | 10 - 11 | 586 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
