Assay Details
Gene Symbol: VPS2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100276.4 | NP_563696.1 | 9 - 10 | 682 | 77 | |
GenBank mRNA | AY052238.1 | - | 9 - 10 | 677 | 77 | |
AY060501.1 | - | 9 - 10 | 529 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
