Assay Details
Gene Symbol: RPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100308.4 | NP_171923.1 | 3 - 4 | 382 | 69 | |
GenBank mRNA | AY048221.1 | - | 3 - 4 | 303 | 69 | |
AY091703.1 | - | 2 - 3 | 212 | 69 | ||
Z23161.1 | - | 241 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
