Assay Details
| Gene Symbol: HCAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100341.4 | NP_171956.2 | 7 - 8 | 720 | 80 | |
| GenBank mRNA | AK117982.1 | - | 7 - 8 | 661 | 80 | |
| BT002029.1 | - | 7 - 8 | 685 | 80 | ||
| BT006603.1 | - | 7 - 8 | 593 | 80 | ||
More Information
Set Membership: |
|

HCAR
Genomic Map