Assay Details
| Gene Symbol: AT1G04680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100348.4 | NP_563715.1 | 1 - 2 | 296 | 66 | |
| GenBank mRNA | AY065034.1 | - | 1 - 2 | 186 | 66 | |
More Information
Set Membership: |
|

AT1G04680
Genomic Map