Assay Details
| Gene Symbol: RCI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100405.4 | NP_172018.1 | 1 - 2 | 270 | 87 | |
| GenBank mRNA | AK227859.1 | - | 1 - 2 | 265 | 87 | |
| BT004817.1 | - | 1 - 2 | 210 | 87 | ||
| U97684.1 | - | 1 - 2 | 244 | 87 | ||
More Information
Set Membership: |
|

RCI3
Genomic Map