Assay Details
| Gene Symbol: RCI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100405.4 | NP_172018.1 | 3 - 4 | 627 | 68 | |
| GenBank mRNA | AK227859.1 | - | 3 - 4 | 622 | 68 | |
| BT004817.1 | - | 3 - 4 | 567 | 68 | ||
| U97684.1 | - | 3 - 4 | 601 | 68 | ||
More Information
Set Membership: |
|

RCI3
Genomic Map