Assay Details
| Gene Symbol: AT1G05520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100431.2 | NP_563741.1 | 10 - 11 | 2416 | 86 | |
| GenBank mRNA | BT002235.1 | - | 10 - 11 | 2101 | 86 | |
More Information
Set Membership: |
|

AT1G05520
Genomic Map