Assay Details
Gene Symbol: ATX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083995.4 | NP_001077464.4 | 11 - 12 | 2113 | 125 | |
NM_001331587.1 | NP_001321563.1 | 11 - 12 | 2227 | 125 | ||
NM_001331588.1 | NP_001321564.1 | 11 - 12 | 2227 | 125 | ||
NM_100464.7 | NP_172074.6 | 11 - 12 | 2197 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
