Assay Details
Gene Symbol: ATX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083995.4 | NP_001077464.4 | 23 - 24 | 3314 | 93 | |
NM_001331587.1 | NP_001321563.1 | 23 - 24 | 3428 | 93 | ||
NM_001331588.1 | NP_001321564.1 | 23 - 24 | 3428 | 93 | ||
NM_100464.7 | NP_172074.6 | 23 - 24 | 3398 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
