Assay Details
Gene Symbol: ATX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083995.4 | NP_001077464.4 | 8 - 9 | 1716 | 90 | |
NM_001331587.1 | NP_001321563.1 | 8 - 9 | 1830 | 90 | ||
NM_001331588.1 | NP_001321564.1 | 8 - 9 | 1830 | 90 | ||
NM_100464.7 | NP_172074.6 | 8 - 9 | 1800 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
