Assay Details
Gene Symbol: ACD32.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100526.5 | NP_172134.1 | 4 - 5 | 786 | 80 | |
GenBank mRNA | BT025652.1 | - | 4 - 5 | 544 | 80 | |
DQ403189.1 | - | 4 - 5 | 544 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
