Assay Details
| Gene Symbol: AT1G06550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100534.4 | NP_172142.2 | 15 - 16 | 1326 | 96 | |
| GenBank mRNA | AK221578.1 | - | 15 - 16 | 1170 | 96 | |
| BT022051.1 | - | 14 - 15 | 1128 | 96 | ||
| BT026035.1 | - | 14 - 15 | 1113 | 96 | ||
More Information
Set Membership: |
|

AT1G06550
Genomic Map