Assay Details
| Gene Symbol: AT1G06550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100534.4 | NP_172142.2 | 4 - 5 | 396 | 65 | |
| GenBank mRNA | AK221578.1 | - | 4 - 5 | 240 | 65 | |
| BT022051.1 | - | 3 - 4 | 198 | 65 | ||
| BT026035.1 | - | 3 - 4 | 183 | 65 | ||
More Information
Set Membership: |
|

AT1G06550
Genomic Map