Assay Details
| Gene Symbol: AT1G06590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100538.5 | NP_172146.2 | 13 - 14 | 2094 | 143 | |
| GenBank mRNA | AY138229.1 | - | 13 - 14 | 1850 | 143 | |
| Gene Symbol: POR C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK226626.1 | - | 1511 | 143 | ||
More Information
Set Membership: |
|

AT1G06590
Genomic Map