Assay Details
| Gene Symbol: AT1G06590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100538.5 | NP_172146.2 | 18 - 19 | 2777 | 105 | |
| GenBank mRNA | AY138229.1 | - | 18 - 19 | 2533 | 105 | |
| Gene Symbol: POR C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK226626.1 | - | 2194 | 105 | ||
More Information
Set Membership: |
|

AT1G06590
Genomic Map