Assay Details
| Gene Symbol: CRTISO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331668.1 | NP_001322208.1 | 3 - 4 | 1044 | 122 | |
| NM_001331669.1 | NP_001322209.1 | 3 - 4 | 1044 | 122 | ||
| NM_001331670.1 | NP_001322210.1 | 3 - 4 | 1024 | 122 | ||
| NM_100559.4 | NP_172167.2 | 3 - 4 | 1283 | 122 | ||
More Information
Set Membership: |
|

CRTISO
Genomic Map