Assay Details
| Gene Symbol: F2KP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100584.5 | NP_172191.1 | 4 - 5 | 956 | 95 | |
| GenBank mRNA | AB035288.1 | - | 4 - 5 | 896 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F2KP| Gene Symbol: F2KP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100584.5 | NP_172191.1 | 4 - 5 | 956 | 95 | |
| GenBank mRNA | AB035288.1 | - | 4 - 5 | 896 | 95 | |
Set Membership: |
|