Assay Details
| Gene Symbol: UBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112954.4 | NP_566666.2 | 1 - 2 | 300 | 89 | |
| GenBank mRNA | AK226894.1 | - | 1 - 2 | 150 | 89 | |
| AX765820.1 | - | 1 - 2 | 93 | 89 | ||
| AY140096.1 | - | 1 - 2 | 155 | 89 | ||
More Information
Set Membership: |
|

UBP14
Genomic Map