Assay Details
| Gene Symbol: UBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112954.4 | NP_566666.2 | 3 - 4 | 595 | 66 | |
| GenBank mRNA | AK226894.1 | - | 3 - 4 | 445 | 66 | |
| AX765820.1 | - | 3 - 4 | 388 | 66 | ||
| AY140096.1 | - | 3 - 4 | 450 | 66 | ||
More Information
Set Membership: |
|

UBP14
Genomic Map