Assay Details
| Gene Symbol: UCNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112974.3 | NP_188719.1 | 1 | 1344 | 97 | |
| GenBank mRNA | BT026453.1 | - | 1 | 1152 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UCNL| Gene Symbol: UCNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112974.3 | NP_188719.1 | 1 | 1344 | 97 | |
| GenBank mRNA | BT026453.1 | - | 1 | 1152 | 97 | |
Set Membership: |
|