Assay Details
Gene Symbol: NF-YA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112983.5 | NP_566670.1 | 3 - 4 | 499 | 89 | |
GenBank mRNA | AF412056.1 | - | 3 - 4 | 501 | 89 | |
AY087696.1 | - | 3 - 4 | 503 | 89 | ||
AY093971.1 | - | 2 - 3 | 359 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
