Assay Details
Gene Symbol: NF-YA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112983.5 | NP_566670.1 | 5 - 6 | 729 | 85 | |
GenBank mRNA | AF412056.1 | - | 5 - 6 | 731 | 85 | |
AY087696.1 | - | 5 - 6 | 733 | 85 | ||
AY093971.1 | - | 4 - 5 | 589 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
